1,2-Dichloro-4-fluoro-5-nitrobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4,5-Dichloro-2-fluoronitrobenzene

D4694-5G D4694-25G
TCIAD4694-5GEA 630 SEK
TCIAD4694-5G TCIAD4694-25G
1,2-Dichloro-4-fluoro-5-nitrobenzene ≥98.0%
1,2-Dichloro-4-fluoro-5-nitrobenzene
Formel: C₆H₂Cl₂FNO₂
MW: 209,99 g/mol
Kokpunkt: 247°C
Smältpunkt: 17°C
MDL Number: MFCD00075330
CAS nummer: 2339-78-8

Order Now

Specifikation testresultat

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR