1,3-Dibromo-5-iodobenzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D4691-1G
D4691-5G
TCIAD4691-1GEA
565
SEK
TCIAD4691-1G
TCIAD4691-5G
1,3-Dibromo-5-iodobenzene ≥98.0%
1,3-Dibromo-5-iodobenzene
Formel:
C₆H₃Br₂I MW: 361,80 g/mol |
MDL Number:
MFCD07778996 CAS nummer: 19752-57-9 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...