Dipropargylamine ≥97.0%

Leverantör: TCI
Fara

Synonyms: Di(2-propynyl)amine

D4685-1ML D4685-5ML
TCIAD4685-1MLEA 950 SEK
TCIAD4685-1ML TCIAD4685-5ML
Dipropargylamine ≥97.0%
Dipropargylamine
Formel: C₆H₇N
Kokpunkt: 65°C
Flampunkt: 46°C
MDL Number: MFCD00008574
CAS nummer: 6921-28-4
UN: 2734
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Orange to Brown clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR