N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan) ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 6-Propionyl-2-dimethylaminonaphthalene, 1-(6-(Dimethylamino)naphthalen-2-yl)propan-1-one, Prodan, 2-(Dimethylamino)-6-propionylnaphthalene

D4678-200MG D4678-1G
TCIAD4678-200MGEA 1250 SEK
TCIAD4678-200MG TCIAD4678-1G
N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan) ≥98.0%
N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan)
Formel: C₁₅H₁₇NO
MW: 227,302 g/mol
MDL Number: MFCD00056615
CAS nummer: 70504-01-7

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR