2,6-Di(1-pyrazolyl)pyridine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D4672-1G
D4672-5G
TCIAD4672-1GEA
1325
SEK
TCIAD4672-1G
TCIAD4672-5G
2,6-Di(1-pyrazolyl)pyridine ≥98.0%
2,6-Di(1-pyrazolyl)pyridine
Formel:
C₁₁H₉N₅ MW: 211,23 g/mol |
MDL Number:
MFCD28336856 CAS nummer: 123640-38-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 137.0 to 141.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...