1,5-Dibromonaphthalene ≥98.0%
Leverantör: TCI EUROPE
Varning
D4660-1G D4660-5G
TCIAD4660-1GEA 1125 SEK
TCIAD4660-1G TCIAD4660-5G
1,5-Dibromonaphthalene ≥98.0%
1,5-Dibromonaphthalene
Formel: C₁₀H₆Br₂
MW: 285,9600 g/mol
Kokpunkt: 326°C
MDL Number: MFCD00089984
CAS nummer: 7351-74-8
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 132.0 deg-C