4,6-Dichloroquinoline ≥98.0%

Leverantör: TCI EUROPE
Varning

D4627-1G D4627-5G
TCIAD4627-1GEA 1200 SEK
TCIAD4627-1G TCIAD4627-5G
4,6-Dichloroquinoline ≥98.0%
4,6-Dichloroquinoline
Formel: C₉H₅Cl₂N
MW: 198,05 g/mol
MDL Number: MFCD00156141
CAS nummer: 4203-18-3

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C
melting range max. 5.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR