4,7-Dibromo-2,1,3-benzoxadiazole ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
4,7-Dibromobenzo[c][1,2,5]oxadiazole, 4,7-Dibromobenzofurazan
D4614-200MG
D4614-1G
TCIAD4614-200MGEA
925
SEK
TCIAD4614-200MG
TCIAD4614-1G
4,7-Dibromo-2,1,3-benzoxadiazole ≥98.0%
4,7-Dibromo-2,1,3-benzoxadiazole
Formel:
C₆H₂Br₂N₂O MW: 277,90 g/mol |
MDL Number:
MFCD02114623 CAS nummer: 54286-63-4 |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 111.0 to 115.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...