9,9-Dimethyl-2-nitrofluorene ≥98.0%

Leverantör: TCI

Synonyms: 9,9-Dimethyl-2-nitro-9H-fluorene

D4609-5G D4609-25G
TCIAD4609-5GEA 445 SEK
TCIAD4609-5G TCIAD4609-25G
9,9-Dimethyl-2-nitrofluorene ≥98.0%
9,9-Dimethyl-2-nitrofluorene
Formel: C₁₅H₁₃NO₂
MW: 239.27 g/mol
MDL Number: MFCD17170053
CAS nummer: 605644-46-0

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR