4,6-Dibromopyrimidine ≥98.0%
Leverantör: TCI
|
Varning
|
D4600-1G
D4600-5G
TCIAD4600-1GEA
630
SEK
TCIAD4600-1G
TCIAD4600-5G
4,6-Dibromopyrimidine ≥98.0%
4,6-Dibromopyrimidine
Formel:
C₄H₂Br₂N₂ |
MDL Number:
MFCD00233946 CAS nummer: 36847-10-6 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 50.0 to 54.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...