Diethyl (chloromethyl)phosphonate ≥98.0%

Leverantör: TCI
Varning

Synonyms: (Chloromethyl)phosphonic Acid Diethyl Ester

D4588-1G D4588-5G
TCIAD4588-1GEA 1350 SEK
TCIAD4588-1G TCIAD4588-5G
Diethyl (chloromethyl)phosphonate ≥98.0%
Diethyl (chloromethyl)phosphonate
Formel: C₅H₁₂ClO₃P
Kokpunkt: 110°C
Flampunkt: 86°C
MDL Number: MFCD00010189
CAS nummer: 3167-63-3
EINECS: 221-632-5

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR