2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-(4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl)benzene-1,3-diol

D4527-5G D4527-25G
TCIAD4527-5GEA 845 SEK
TCIAD4527-5G TCIAD4527-25G
2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine
Formel: C₂₅H₂₃N₃O₂
MW: 397,48 g/mol
MDL Number: MFCD16251125
CAS nummer: 1668-53-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 203.0 to 207.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR