Bis(p-tolyl)phosphine oxide ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Di-p-tolylphosphine oxide, Di(p-tolyl)phosphine Oxide

D4520-1G D4520-5G
TCIAD4520-1GEA 775 SEK
TCIAD4520-1G TCIAD4520-5G
Bis(p-tolyl)phosphine oxide ≥95.0%
Bis(p-tolyl)phosphine oxide
Formel: C₁₄H₁₅OP
MW: 230,24 g/mol
Smältpunkt: 96°C
MDL Number: MFCD01445489
CAS nummer: 2409-61-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR