2,4-Dimethylbenzonitrile ≥98.0%

Leverantör: TCI EUROPE
Varning

D4495-5G D4495-25G
TCIAD4495-5GEA 815 SEK
TCIAD4495-5G TCIAD4495-25G
2,4-Dimethylbenzonitrile ≥98.0%
2,4-Dimethylbenzonitrile
Formel: C₉H₉N
MW: 131,18 g/mol
Kokpunkt: 109°C
MDL Number: MFCD00016377
CAS nummer: 21789-36-6
EINECS: 244-580-5

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR