1,1,1,2,2,3,4,5,5,5-Decafluoro-3-methoxy-4-(trifluoromethyl)pentane ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
3-Methoxyperfluoro(2-methylpentane)
D4484-25G
D4484-500G
TCIAD4484-25GEA
375
SEK
TCIAD4484-25G
TCIAD4484-500G
1,1,1,2,2,3,4,5,5,5-Decafluoro-3-methoxy-4-(trifluoromethyl)pentane ≥98.0%
1,1,1,2,2,3,4,5,5,5-Decafluoro-3-methoxy-4-(trifluoromethyl)pentane
Formel:
C₇H₃F₁₃O MW: 350,08 g/mol Kokpunkt: 100°C |
MDL Number:
MFCD07784227 CAS nummer: 132182-92-4 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...