7H-Dibenzo[c,g]carbazole ≥98.0%

Leverantör: TCI EUROPE
Varning

D4473-200MG D4473-1G
TCIAD4473-200MGEA 1325 SEK
TCIAD4473-200MG TCIAD4473-1G
7H-Dibenzo[c,g]carbazole ≥98.0%
7H-Dibenzo[c,g]carbazole
Formel: C₂₀H₁₃N
MW: 267,33 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00215941
CAS nummer: 194-59-2
EINECS: 205-895-3

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR