Difluoromethyl-2,2,3,3-tetrafluoropropyl ether ≥98.0%
Leverantör: TCI EUROPE
Fara
D4472-1G D4472-5G
TCIAD4472-1GEA 695 SEK
TCIAD4472-1G TCIAD4472-5G
Difluoromethyl-2,2,3,3-tetrafluoropropyl ether ≥98.0%
Difluoromethyl-2,2,3,3-tetrafluoropropyl ether
Formel: C₄H₄F₆O
MW: 182,07 g/mol
Kokpunkt: 76°C
MDL Number: MFCD00236119
CAS nummer: 35042-99-0
UN: 3271
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %