Dimethoxypillar[5]arene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 12,15,32,35,52,55,72,75,92,95-Decamethoxy-1,3,5,7,9(1,4)-pentabenzenacyclodecaphane, DMpillar[5]arene

D4471-100MG D4471-1G
TCIAD4471-100MGEA 615 SEK
TCIAD4471-100MG TCIAD4471-1G
Dimethoxypillar[5]arene ≥98.0%
Dimethoxypillar[5]arene
Formel: C₄₅H₅₀O₁₀
MW: 750,89 g/mol
Smältpunkt: 249°C
MDL Number: MFCD28386103
CAS nummer: 1188423-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2252bc55ca410853d0e137509116ccdb9484e7c2&vid=2252bc55ca410853d0e137509116ccdb9484e7c2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR