3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)] ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: DiSC2(3), 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzothiazolylidene]-1-propenyl]benzothiazolium Iodide, 3,3-Diethylthiacarbocyanine iodide, 3-Ethyl-2-(3-(3-ethylbenzo[d]thiazol-2(3H)-ylidene)prop-1-en-1-yl)benzo[d]thiazol-3-ium iodide

D4454-1G D4454-5G
TCIAD4454-1GEA 1625 SEK
TCIAD4454-1G TCIAD4454-5G
3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)] ≥98.0% (by HPLC, total nitrogen)
3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)]
Formel: C₂₁H₂₁IN₂S₂
MW: 492,44 g/mol
Smältpunkt: 268°C
MDL Number: MFCD00011956
CAS nummer: 905-97-5

Order Now

Specifikation testresultat

Appearance Amber to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Lambda max. 555.0 to 560.0 nm (MeOH)
Absorbance (E1%1cm) min. 3000 (MeOH, 555.0 to 560.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR