2,4-Dichloroquinoline ≥98.0%
Leverantör: TCI EUROPE
Varning
D4452-5G D4452-25G
TCIAD4452-5GEA 1225 SEK
TCIAD4452-5G TCIAD4452-25G
2,4-Dichloroquinoline ≥98.0%
2,4-Dichloroquinoline
Formel: C₉H₅Cl₂N
MW: 198,0500 g/mol
Kokpunkt: 282°C
MDL Number: MFCD00023939
CAS nummer: 703-61-7
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C