Dichlorobis(2-ethylhexyl)silane ≥95.0%
Leverantör: TCI
|
Fara
|
D4443-1G
D4443-5G
TCIAD4443-1GEA
725
SEK
TCIAD4443-1G
TCIAD4443-5G
Dichlorobis(2-ethylhexyl)silane ≥95.0%
Dichlorobis(2-ethylhexyl)silane
Formel:
C₁₆H₃₄Cl₂Si |
MDL Number:
MFCD22370170 CAS nummer: 1089687-03-5 UN: 2987 ADR: 8,II |
Specifikation testresultat
Appearance | White to Orange to Green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...