4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
4,7-Dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine
D4442-200MG
D4442-1G
TCIAD4442-200MGEA
630
SEK
TCIAD4442-200MG
TCIAD4442-1G
4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine ≥97.0%
4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine
Formel:
C₅HBr₂N₃S MW: 294,95 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD23703147 CAS nummer: 333432-27-2 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 146.0 to 150.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...