4,4'-Dichlorobenzil ≥98.0%
Leverantör: TCI
Varning
Synonyms: 4,4'-Dichlorodibenzoyl, 4,4'-Dichlorobibenzoyl, 1,2-Bis(4-chlorophenyl)ethane-1,2-dione
D4440-1G D4440-5G
TCIAD4440-1GEA 950 SEK
TCIAD4440-1G TCIAD4440-5G
4,4'-Dichlorobenzil ≥98.0%
4,4'-Dichlorobenzil
Formel: C₁₄H₈Cl₂O₂
MW: 279.12 g/mol
Kokpunkt: 220°C
MDL Number: MFCD00018685
CAS nummer: 3457-46-3
EINECS: 222-387-7
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 197.0 to 201.0 deg-C