4,4'-Dimethyl-2,2'-bipiperidine (mixture of isomers) ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

D4435-1G D4435-5G
TCIAD4435-1GEA 1850 SEK
TCIAD4435-1G TCIAD4435-5G
4,4'-Dimethyl-2,2'-bipiperidine (mixture of isomers) ≥98.0% (by GC, titration analysis)
4,4'-Dimethyl-2,2'-bipiperidine (mixture of isomers)
Formel: C₁₂H₂₄N₂
MW: 196,34 g/mol
Kokpunkt: 75°C
CAS nummer: 1378896-71-9

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomer)
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR