2,6-Dichloropyridine-4-methanol ≥98.0%

Leverantör: TCI

Synonyms: 2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine

D4423-1G D4423-5G
TCIAD4423-1GEA 735 SEK
TCIAD4423-1G TCIAD4423-5G
2,6-Dichloropyridine-4-methanol ≥98.0%
2,6-Dichloropyridine-4-methanol
Formel: C₆H₅Cl₂NO
MW: 178.0160 g/mol
MDL Number: MFCD00052638
CAS nummer: 101990-69-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR