2,3-Dichloro-5-nitropyridine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D4416-1G
D4416-5G
TCIAD4416-1GEA
390
SEK
TCIAD4416-1G
TCIAD4416-5G
2,3-Dichloro-5-nitropyridine ≥98.0%
2,3-Dichloro-5-nitropyridine
Formel:
C₅H₂Cl₂N₂O₂ Kokpunkt: 256°C |
MDL Number:
MFCD03840432 CAS nummer: 22353-40-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...