(Difluoromethyl)trimethylsilane ≥97.0%

Leverantör: TCI EUROPE
Fara

D4364-5ML D4364-1ML
TCIAD4364-5MLEA 965 SEK
TCIAD4364-5ML TCIAD4364-1ML
(Difluoromethyl)trimethylsilane ≥97.0%
(Difluoromethyl)trimethylsilane
Formel: C₄H₁₀F₂Si
MW: 124,21 g/mol
Kokpunkt: 52°C
Storage Temperature: Kylskåp
MDL Number: MFCD19381658
CAS nummer: 65864-64-4
UN: 1993
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR