2,7-Dibromophenanthrene-9,10-dione ≥97.0%
Leverantör: TCI EUROPE
Varning
D4363-1G D4363-5G
TCIAD4363-1GEA 735 SEK
TCIAD4363-1G TCIAD4363-5G
2,7-Dibromophenanthrene-9,10-dione ≥97.0%
2,7-Dibromophenanthrene-9,10-dione
Formel: C₁₄H₆Br₂O₂
MW: 366,01 g/mol
Smältpunkt: 331°C
MDL Number: MFCD03931078
CAS nummer: 84405-44-7
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %