(3-(Diethylamino)propyl)trimethoxysilane ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Trimethoxy(3-(diethylamino)propyl)silane, N,N-Diethyl-3-(trimethoxysilyl)propan-1-amine, (3-Diethylaminopropyl)trimethoxysilane, [3-(Diethylamino)propyl]trimethoxysilane, N,N-Diethyl-3-(trimethoxysilyl)propylamine
D4362-5ML
D4362-25ML
TCIAD4362-5MLEA
470
SEK
TCIAD4362-5ML
TCIAD4362-25ML
(3-(Diethylamino)propyl)trimethoxysilane ≥97.0% (by GC, titration analysis)
(3-(Diethylamino)propyl)trimethoxysilane
Formel:
(H₃CCH₂)₂NCH₂CH₂CH₂Si(OCH₃)₃ MW: 235,4 g/mol Flampunkt: 100°C |
MDL Number:
MFCD00039882 CAS nummer: 41051-80-3 EINECS: 255-192-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...