2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0%

Leverantör: TCI EUROPE

Synonyms: (Dimethylphenylsilyl)boronic Acid Pinacol Ester

D4357-1G D4357-5G
TCIAD4357-1GEA 1225 SEK
TCIAD4357-1G TCIAD4357-5G
2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0%
2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₁₄H₂₃BO₂Si
MW: 262,23 g/mol
Kokpunkt: 120°C
MDL Number: MFCD05664111
CAS nummer: 185990-03-8

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR