Dobutamine hydrochloride ≥98.0%

Leverantör: TCI
Varning

D4335-20MG D4335-100MG
TCIAD4335-20MGEA 605 SEK
TCIAD4335-20MG TCIAD4335-100MG
Dobutamine hydrochloride ≥98.0%
Dobutamine hydrochloride
Formel: C₁₈H₂₃NO₃
MDL Number: MFCD00153795
CAS nummer: 49745-95-1
EINECS: 256-464-1
Merck Index: 14,03395

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 4.00 to 4.30 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR