2,5-Dibromopyrazine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D4323-1G
D4323-5G
TCIAD4323-1GEA
335
SEK
TCIAD4323-1G
TCIAD4323-5G
2,5-Dibromopyrazine ≥98.0%
2,5-Dibromopyrazine
Formel:
C₄H₂Br₂N₂ MW: 237,8800 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD08061601 CAS nummer: 23229-26-7 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...