(2,4-Difluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2,4-Difluorophenylacetic acid

D4321-5G D4321-25G
TCIAD4321-5GEA 845 SEK
TCIAD4321-5G TCIAD4321-25G
(2,4-Difluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2,4-Difluorophenyl)acetic acid
Formel: (C₆H₃F₂)CH₂COOH
MW: 172,13 g/mol
MDL Number: MFCD00009999
CAS nummer: 81228-09-3
EINECS: 279-709-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR