2,3-Difluorobenzyl bromide ≥98.0%

Leverantör: TCI
Fara

Synonyms: α-Bromo-2,3-difluorotoluene

D4320-1G D4320-5G
TCIAD4320-1GEA 245 SEK
TCIAD4320-1G TCIAD4320-5G
2,3-Difluorobenzyl bromide ≥98.0%
2,3-Difluorobenzyl bromide
Formel: C₇H₅BrF₂
Flampunkt: 90°C
MDL Number: MFCD00042488
CAS nummer: 113211-94-2
UN: 3265
ADR: 8,II

Order Now

Specifikation testresultat

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR