7,7-Dichlorobicyclo[4.1.0]heptane ≥98.0%
Leverantör: TCI EUROPE
Varning
D4314-5G D4314-25G
TCIAD4314-5GEA 510 SEK
TCIAD4314-5G TCIAD4314-25G
7,7-Dichlorobicyclo[4.1.0]heptane ≥98.0%
7,7-Dichlorobicyclo[4.1.0]heptane
Formel: C₇H₁₀Cl₂
MW: 165,06 g/mol
Kokpunkt: 80°C
MDL Number: MFCD00466186
CAS nummer: 823-69-8
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %