9,9-Dimethyl-9H-9-silafluorene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 5,5-Dimethyl-5H-dibenzo[b,d]silole

D4308-1G D4308-5G
TCIAD4308-1GEA 1425 SEK
TCIAD4308-1G TCIAD4308-5G
9,9-Dimethyl-9H-9-silafluorene ≥98.0%
9,9-Dimethyl-9H-9-silafluorene
Formel: C₁₄H₁₄Si
MW: 210,35 g/mol
MDL Number: MFCD00093801
CAS nummer: 13688-68-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR