2,5-Dibromo-3-methylthiophene ≥98.0%

Leverantör: TCI EUROPE
Varning

D4304-5G D4304-25G
TCIAD4304-5GEA 325 SEK
TCIAD4304-5G TCIAD4304-25G
2,5-Dibromo-3-methylthiophene ≥98.0%
2,5-Dibromo-3-methylthiophene
Formel: C₅H₄Br₂S
MW: 255,97 g/mol
Kokpunkt: 108°C
Storage Temperature: Kylskåp
MDL Number: MFCD00015470
CAS nummer: 13191-36-1
EINECS: 236-147-4

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.9880 to 1.9980

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR