Doxofylline ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 7-(1,3-Dioxolan-2-ylmethyl)theophylline, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethylxanthine, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethyl-1H-purine-2,6-dione

D4302-1G D4302-25G
TCIAD4302-1GEA 325 SEK
TCIAD4302-1G TCIAD4302-25G
Doxofylline ≥98.0% (by HPLC, titration analysis)
Doxofylline
Formel: C₁₁H₁₄N₄O₄
MW: 266,26 g/mol
MDL Number: MFCD00865218
CAS nummer: 69975-86-6
EINECS: 274-239-6
Merck Index: 14,03438

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR