Acetone oxime 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Propan-2-one oxime
102391000 102395000 102390025
ACRO102391000EA 1050 SEK
ACRO102391000 ACRO102395000 ACRO102390025
Acetone oxime 98%
Acetone oxime
Formel: C₃H₇NO
MW: 73,09 g/mol
Kokpunkt: 135 °C (760 mmHg)
Smältpunkt: 58…62 °C
Densitet: 1,045 g/cm³ (20 °C)
Flampunkt: 60 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002118
CAS nummer: 127-06-0
EINECS: 204-820-1
UN: 1325
ADR: 4.1,II
Merck Index: 13,00077
Order Now
Specifikation testresultat
Appearance White to yellow Crystals or crystalline solid
Infrared spectrum Conforms
Melting point 58 to 63 °C
GC ≥97.5 %