2,4-Dibutoxyphenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: (2,4-Dibutoxyphenyl)boronic acid

D4263-1G D4263-5G
TCIAD4263-1GEA 3550 SEK
TCIAD4263-1G TCIAD4263-5G
2,4-Dibutoxyphenylboronic acid (contains varying amounts of Anhydride)
2,4-Dibutoxyphenylboronic acid
Formel: C₁₄H₂₃BO₄
MW: 266,14 g/mol
MDL Number: MFCD07784348
CAS nummer: 870778-89-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR