1-(2,6-Dichlorophenyl)oxindole ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1-(2,6-Dichlorophenyl)-2-indolinone, 1-(2,6-Dichlorophenyl)-2-oxoindoline, Diclofenac Amide

D4260-5G D4260-25G
TCIAD4260-5GEA 310 SEK
TCIAD4260-5G TCIAD4260-25G
1-(2,6-Dichlorophenyl)oxindole ≥98.0%
1-(2,6-Dichlorophenyl)oxindole
Formel: C₁₄H₉Cl₂NO
MW: 278,13 g/mol
MDL Number: MFCD00451393
CAS nummer: 15362-40-0
EINECS: 239-399-3

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR