10,10'-Dibromo-9,9'-bianthracene ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
D4254-1G
D4254-5G
TCIAD4254-1GEA
1650
SEK
TCIAD4254-1G
TCIAD4254-5G
10,10'-Dibromo-9,9'-bianthracene ≥98.0% (by HPLC, titration analysis)
10,10'-Dibromo-9,9'-bianthracene
Formel:
C₂₈H₁₆Br₂ MW: 512,24 g/mol Smältpunkt: 342°C |
MDL Number:
MFCD11045035 CAS nummer: 121848-75-7 |
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...