6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic acid ≥97.0% (by HPLC, titration analysis)
Leverantör: TCI
|
Fara
|
D4242-1G
D4242-5G
TCIAD4242-1GEA
605
SEK
TCIAD4242-1G
TCIAD4242-5G
6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic acid ≥97.0% (by HPLC, titration analysis)
6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic acid
Formel:
C₁₆H₁₂O₄ |
MDL Number:
MFCD00242952 CAS nummer: 55453-87-7 UN: 2811 ADR: 6.1,III Merck Index: 14,05237 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 140.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...