2-(Dimethylamino)-2-oxoacetic acid ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
N,N-Dimethyloxamic acid, N,N-Dimethyloxamidic acid
D4241-5G
D4241-1G
TCIAD4241-5GEA
895
SEK
TCIAD4241-5G
TCIAD4241-1G
2-(Dimethylamino)-2-oxoacetic acid ≥97.0%
2-(Dimethylamino)-2-oxoacetic acid
Formel:
C₄H₇NO₃ MW: 117,1000 g/mol |
MDL Number:
MFCD01971023 CAS nummer: 32833-96-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...