2,5-Dibromopyrimidine ≥98.0%

Leverantör: TCI EUROPE
Varning

D4225-1G D4225-5G
TCIAD4225-1GEA 725 SEK
TCIAD4225-1G TCIAD4225-5G
2,5-Dibromopyrimidine ≥98.0%
2,5-Dibromopyrimidine
Formel: C₄H₂Br₂N₂
MW: 237,8800 g/mol
MDL Number: MFCD08275684
CAS nummer: 32779-37-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR