3,4-Dihexylthiophene ≥97.0%

Leverantör: TCI

D4202-1G D4202-5G
TCIAD4202-1GEA 815 SEK
TCIAD4202-1G TCIAD4202-5G
3,4-Dihexylthiophene ≥97.0%
3,4-Dihexylthiophene
Formel: C₁₆H₂₈S
Kokpunkt: 103°C
MDL Number: MFCD21363049
CAS nummer: 122107-04-4

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR