Doxorubicin hydrochloride ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Doxorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-10-(((2R,4S,5S,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride
D4193-25MG
D4193-100MG
TCIAD4193-25MGEA
710
SEK
TCIAD4193-25MG
TCIAD4193-100MG
Doxorubicin hydrochloride ≥95.0%
Doxorubicin hydrochloride
Formel:
C₂₇H₂₉NO₁₁·HCl MW: 579,98 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00077757 CAS nummer: 25316-40-9 EINECS: 246-818-3 Merck Index: 14,03439 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Specific rotation [a]20/D | +240 to +270 deg (C=0.1,MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...