5,5'-Dibromo-4,4'-dihexyl-2,2'-bithiophene ≥95.0%
Leverantör: TCI
D4184-200MG
D4184-1G
TCIAD4184-200MGEA
1175
SEK
TCIAD4184-200MG
TCIAD4184-1G
5,5'-Dibromo-4,4'-dihexyl-2,2'-bithiophene ≥95.0%
5,5'-Dibromo-4,4'-dihexyl-2,2'-bithiophene
Formel:
C₂₀H₂₈Br₂S₂ Storage Temperature: Kylskåp |
CAS nummer:
214493-03-5 |
Specifikation testresultat
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...