Etidronate disodium hydrate ≥98.0%

Leverantör: TCI
Varning

D4159-5G D4159-25G
TCIAD4159-5GEA 790 SEK
TCIAD4159-5G TCIAD4159-25G
Etidronate disodium hydrate ≥98.0%
Etidronate disodium hydrate
Formel: C₂H₆Na₂O₇P₂·H₂O
Storage Temperature: Kylskåp
MDL Number: MFCD00152567
CAS nummer: 7414-83-7
EINECS: 231-025-7
Merck Index: 14,03863
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e761632b19d1153069f2d5fd5cdfce2a22dbc30a&vid=e5264051bdb6d234170d1a3f2f8e8455ddfd6bb2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 % (after drying)
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR