2,4-Dichloro-5-iodopyrimidine ≥98.0%

Leverantör: TCI EUROPE
Varning

D4122-1G D4122-5G
TCIAD4122-1GEA 390 SEK
TCIAD4122-1G TCIAD4122-5G
2,4-Dichloro-5-iodopyrimidine ≥98.0%
2,4-Dichloro-5-iodopyrimidine
Formel: C₄HCl₂IN₂
MW: 274,87 g/mol
MDL Number: MFCD01898087
CAS nummer: 13544-44-0

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR